Dräger™ CMS Chips: Methylene Chloride
Suitable for 10 measurements of one gas
Supplier:  Dräger™ 6406510
Catalog No. 19-046204
Explore available promotions
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
1 to 30mg/L
180 to 600 sec.
10° to 40°C
Methylene Chloride CMS Chip
20 to 400
50°F to 104°F