missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dräger™ CMS Chips: Phosphine

Suitable for 10 measurements of one gas.

$208.69 - $208.69

Specifications

Air Pressure 700 to 1100 hpa
Type Phosphine CMS Chip
Humidity Range 1 to 30mg/L
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
View More Specs

Products 1
Catalog Number Mfr. No. Type Measurement Range Response Time Price Quantity  
Catalog Number Mfr. No. Type Measurement Range Response Time Price Quantity  
19-041692
View Documents
Dräger™
6406400
Phosphine CMS Chip 0.1 to 2.5 25 to 350 sec.
Each for $208.69
 
Description

Description

Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications

Specifications

700 to 1100 hpa
1 to 30mg/L
0°C to 40°C
Phosphine CMS Chip
32°F to 104°F
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'