Dräger™ Demand Flow Regulators
For use with instruments equipped with sample pumps
Supplier: Dräger™ 4594641
Description
- Calibration gas flows “on-demand” when the instrument or E-Cal module pump is activated
- 0.625 in. -18 UNF thread size
Specifications
Flow Demand Regulator | |
58 and 103 L cylinder, instruments equipped with sample pump, E-Cal system | |
5/8 UNF thread |