Dräger™ Flow Check Air Flow Indicator Kit
Generates a plume of smoke, making the direction and dispersion of air currents visible, as well as their relative speed
Supplier:  Dräger™ 6400761
Catalog No. 19-038773
Explore available promotions
Includes: Three ampules, a nozzle, and rechargeable NiCd batteries that offer about 20 minutes of operation per charge
Description
Description
Ideal for any applications where ventilation controls the health and safety of workers, such as paint spray booths and fume hoods. Also useful tool for air balancing, adjusting ventilation louvers in offices and institutions, and performing indoor air quality studies. Acts as a leak detector by using smoke to check airflow outside airtight doors, detecting leaks into cleanrooms or containment areas.
  • Lightweight, ergonomically designed unit offers easy, one-handed operation
  • Generates environmentally safe smoke in puffs or in a continuous cloud
  • Each smoke-generating ampule provides about five minutes of smoke
Order Info
Measures approximately 11 3/4 x 7 x 2 3/4 in. (300 x 200 x 70mm) and weighs 171/2 oz. (500g). Chargers and replacement ampules are available separately. For charger, see Cat. No. 19-130-1060.
Specifications
Specifications
Air Indicator Kit
Videos
SDS
Documents
Documents
Accessories