Dräger™ Short-Term Detector Tubes: Carbon Monoxide
Detector tubes
$162.77 - $229.73
Specifications
Components Measured Carbon Monoxide
Quantity 10
View More Specs
Products 5
Catalog Number Mfr. No. Components Measured Tube Type Measurement Range Humidity Range Price Quantity  
Catalog Number Mfr. No. Components Measured Tube Type Measurement Range Humidity Range Price Quantity  
17-985-198
View Documents
Dräger™
6733051
Carbon Monoxide Carbon monoxide 2/a 2 to 300 2 to 20mg/L
Pack of 10 Each for $183.36
29-663-361
View Documents
Dräger™
6728511
Carbon Monoxide Carbon monoxide 5/a-P 1:2.5 to 150 0 to 50mg/L
Pack of 10 Each for $204.04
17-985-199
View Documents
Dräger™
CH25601
Carbon Monoxide Carbon monoxide 5/c2 5 to 700 50mg/L
Pack of 10 Each for $183.36
17-985-200
View Documents
Dräger™
CH19701
Carbon Monoxide Carbon monoxide 8/a (only for CO in H2) 8 to 150 Less than50mg/L
Pack of 10 Each for $229.73
17-985-201
View Documents
Dräger™
CH20601
Carbon Monoxide Carbon monoxide 10/b2 10 to 3000 50mg/L
Pack of 10 Each for $162.77
Description
Description
Detector tubes
 Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Specifications
Carbon Monoxide
10
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'