Dräger™ X-plore™ Ammonia/Methylamine (AM/MA) Cartridge
For protection against Ammonia and Methylamine gases
Supplier: Dräger™ 6738027
Description
Fits Dräger X-plore 3300/3500 half masks and Dräger X-plore 5500 full face masks
- Strong blue ABS plastic housing
- Safe, quick and easy bayonet connection with clear guidance bringing the filter/cartridge securely into place
- Both filters are inserted in a top-down position
- Option to upgrade a gas cartridge to a combination cartridge by simply adding a pad retrainer and a N 95 or R 95 pad
- Shelflife (from date of production): gas and combination cartridges 6 years, pads 3 years
- To convert the AM/MA cartridge to a combination cartridge, you will need two filter pad retainers (p/n: 6738038) and two N95 Prefilters (p/n: 6738038).
Order Info
Packaged in pairs for optimum protection of the unused filter. Instructions included with each pair.
Certifications
NIOSH 42 CFR 84
Specifications
Gas/Vapor Cartridge | |
Blue | |
Cartridge | |
1 Ea. |
NIOSH | |
ABS Plastic | |
Mask |