missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DR Instruments Heavy Duty Bone Shears, Stainless Steel, 7 in.

Bone Shears are a necessary tool for a dissection lab. With the help of the bone shears one can prepare / cut bone specimens for further study.

Supplier:  DR Instruments BS70

Catalog No. S08105



Explore available promotions

Description

Description

DR Instruments bone cutters are designed to cut heavy bones without smashing the bone specimens. Super sharp blades provides clear cut every time.

Features:

  • Made of Surgical Stainless-steel
  • Non-slip handle design
  • Features single or double spring settings
  • Hand honed jaws for clean cut.
  • With side cutting action
  • Length: 17.7 cm (7 in.)
Specifications

Specifications

Bone-cutting Forceps
7 in.
Stainless Steel
Stainless Steel
17.7 cm
Videos
SDS
Documents

Documents

Promotions

Promotions