missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DR Instruments Insect Pinning Forceps, Angular Points, Stainless Steel
Insect pinning forceps are a specialized tool used by entomologists and insect collectors to carefully handle and pin insects for preservation and display.
Supplier: DR Instruments 20
Description
DR Instruments insect pinning forceps have been meticulously crafted to cater to the delicate requirements of entomology and insect collection. With their precise points and angled tips, these forceps offer exceptional maneuverability and ease of use.
Key Features:
- Non-slip serrated handles for a secure grip.
- Angled tips for effortless handling.
- Constructed from durable stainless steel for long-lasting performance.
- Serrated tips prevent insects from slipping during handling.
- Compact 4.5 inch length for convenient use in various entomological tasks.
Specifications
Forcep | |
For insect pinning | |
11.4 cm |
Stainless Steel | |
4.5 in. |