missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DR Instruments Mall Probe and Seeker, Stainless Steel, 6 in.
The Mall Probe and Seeker is an important dissection tool for separating and tracking veins, nerves, and organs from connective tissue.
Supplier: DR Instruments 36
Description
- Fabricated with surgical grade stainless steel
- Angular medium point at one end
- Length: 15.24 cm (6 in.)
Specifications
Mall Probe | |
Stainless Steel | |
6 in. | |
15.24 cm |