missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Drabkin's Reagent, Ricca Chemical
Drabkin's Reagent for measurement of total hemoglobin.
Supplier: Ricca Chemical Company 266016
Description
- For total hemoglobin measurement
Specifications
Certificate of Analysis | |
Not Applicable | |
Colorimetric | |
Poly Amber Bottle | |
Drabkin's Reagent | |
500 mL |
No | |
Hemoglobin | |
No | |
Blood | |
Measurement of total hemaglobins | |
Liquid |