missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ DRAQ5™

Anthraquinone dye with high affinity for double-stranded DNA. It is a membrane-permeable dye that can label live or fixed/dead cells.

$373.00 - $898.00

Specifications

Color Red
Detection Method Fluorescence
For Use With (Application) Immunohistochemistry (IHC), Flow Cytometry
For Use With (Equipment) Fluorescence Microscope
Product Type Dye
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016967
View Documents
Invitrogen™
65088092
50μL
Each for $373.00
 
5016968
View Documents
Invitrogen™
65088096
200μL
Each for $898.00
 
Description

Description

  • In flow cytometry, this dye can be used to distinguish nucleated and non-nucleated cells.
  • DRAQ5™ can also be used to report nuclear DNA content for ploidy and cell cycle analysis because it binds DNA stoichiometrically. In fluorescent microscopy, it can be used as a nuclear counterstain
  • Excited by wavelengths from 488 to 647nm
  • For microscopy/imaging, 633 or 647nm sources for excitation are recommended
  • For flow cytometry, with excitation at 488nm, this dye can be detected with filter sets designed for PerCP-eFluor™ 710 or PerCP-Cy5.5 (685LP dichroic mirror and 710/50 band pass); with excitation at 633nm, it can be detected with filter sets designed for APC (660/20 band pass)
  • For cell cycle/DNA analysis applications, a higher wavelength filter is recommended, such as a 710LP or a filter set designed for PE-Cy7 (735LP dichroic mirror and 780/60 band pass), to optimize the CV for the G1 and G2/M peaks
  • Because of its broad excitation and emission properties, DRAQ5™ is not recommended in combination with other far-red fluorochromes excited by the 488 or 633nm laser lines, including PE-Cy7, PerCP-eFluor™ 710, PerCP-Cy5.5, APC, Alexa Fluor™ 647, Alexa Fluor™ 700, APC-eFluor™ 780 and equivalents (APC-Alexa Fluor™ 750 and APC-Cy7)
Specifications

Specifications

Red
Immunohistochemistry (IHC), Flow Cytometry
Dye
eBioscience™
Fluorescence
Fluorescence Microscope
488-647⁄633-710
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.