Supelco Drawer Unit
The Drawer Unit is a spare part suitable for the TLC Explorer Documentation System. The TLC Explorer Documentation System is designed to document and evaluate thin-layer chromatographic (TLC) plates.
Supplier:  Supelco 1526150001
Catalog No. 15-261-50001
Explore available promotions
Description
Description
  • The Drawer unit is a spare part suitable for the TLC Explorer Documentation System.
  • The TLC Explorer Documentation System is designed to document and evaluate thin-layer chromatographic (TLC) plates.
  • It offers a safe, fast and flexible way to visually inspect and digitally record images of TLC plates after their chromatographic development workflow in the laboratory.
  • For detecting different substance classes, the device contains three different LED light sources (white light (VIS), UVA (366 nm) and UVC (254 nm), which illuminate the plates from above in direct-light mode.
  • Recorded plate images can be evaluated to generate reports or export the data for further processing with other systems.
Specifications
Specifications
Drawer Unit