missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase

Thermo Scientific DreamTaq Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases.

$165.00 - $5920.00

Specifications

GC-Rich PCR Performance Low
Polymerase DreamTaq DNA Polymerase
Reaction Speed Standard
For Use With (Application) Hot-start PCR
Concentration 5 U/μL
View More Specs

Products 7
Catalog Number Mfr. No. Color Quantity Price Quantity  
Catalog Number Mfr. No. Color Quantity Price Quantity  
FEREP1701
View Documents
Thermo Scientific™
EP1701
Colorless 200 U
Each for $165.00
 
FEREP1702
View Documents
Thermo Scientific™
EP1702
Colorless 500 U
Each for $376.00
 
FEREP1704
View Documents
Thermo Scientific™
EP1704
Colorless 4 x 2500 U
Each for $5,920.00
 
FEREP1711
View Documents
Thermo Scientific™
EP1711
Green 200 U
Each for $166.00
 
FEREP1712
View Documents
Thermo Scientific™
EP1712
Green 500 U
Each for $376.00
 
FEREP1713
View Documents
Thermo Scientific™
EP1713
Green 2500 U
Each for $1,604.00
 
FEREP1714
View Documents
Thermo Scientific™
EP1714
Green 4 x 2500 U
Each for $5,850.00
 
Description

Description

Thermo Scientific DreamTaq Hot Start DNA Polymerase, available in colorless and green formats, is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases.

DreamTaq Hot Start DNA Polymerase employs antibody-based inhibition of DNA polymerase activity at ambient temperatures to prevent the amplification of non-specific products prior to the amplification step. With DreamTaq Hot Start DNA Polymerase, reactions can be set up at room temperature using the same protocol and cycling conditions as conventional Taq DNA polymerases. The enzyme is supplied with an optimized 10X DreamTaq Buffer containing magnesium chloride, which eliminates the need for extensive optimization of reaction conditions. DreamTaq Hot Start DNA Polymerase is also available with pre-added density reagent and electrophoresis tracking dyes, as well as in master mix format.

The 10X DreamTaq Green Buffer also includes a density reagent and two tracking dyes for convenient direct loading of PCR products on a gel.

DreamTaq Hot Start DNA Polymerase is formulated to enhance productivity through:

  • Better reaction outcomes
    • Higher yield of target amplicons from low template amounts
    • Increased reaction specificity due to reliable hot-start technology
    • Wider range of amplicon lengths with routine amplification of genomic DNA fragments up to 6 kb
  • Enhanced convenience
    • Room temperature reaction setup
    • Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer
    • Direct loading of PCR products on a gel with DreamTaq Green Buffer

Applications

  • Generation of PCR products for TA cloning
  • Routine PCR
  • Colony PCR
  • Genotyping
  • RT-PCR
Specifications

Specifications

Low
Standard
5 U/μL
3'-A
DreamTaq DNA Polymerase
Hot-start PCR
Built-In Hot Start
Separate Components
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'