missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Decon™ Dri-Clean™ Detergent Powder

For manual, soak, or ultrasonic cleaning of all labware

$114.40 - $114.40

Specifications

pH 10.5 ±0.5
Color White
For Use With (Application) For manual, soak, or ultrasonic cleaning, removes Protein, dried Blood, Oil, Grease and most other contaminants from Glassware, Plasticware, Ceramic and Metal
View More Specs

Products 1
Catalog Number Mfr. No. Volume (English) Price Quantity  
Catalog Number Mfr. No. Volume (English) Price Quantity  
04-355-75
View Documents
Decon™
DC2502
4-3/8 lb.
Each for $114.40
Case of 6 Each for $686.40
 
Description

Description

  • Powdered formula
  • Effectively removes protein, dried blood, oil, grease, and most other contaminants from glassware, plasticware, ceramic, and metal
  • Completely free-rinsing, residue-free, and biodegradable
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic

Specifications

Specifications

10.5 ±0.5
White
For manual, soak, or ultrasonic cleaning, removes Protein, dried Blood, Oil, Grease and most other contaminants from Glassware, Plasticware, Ceramic and Metal
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'