missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dried Ferrous Sulfate, Powder, USP, 86-89%, Spectrum™ Chemical

F1065, 13463-43-9, FeSO4·xH2O

$565.18 - $7291.79

Chemical Identifiers

CAS 13463-43-9
Molecular Formula FeH2O5S
Molecular Weight (g/mol) 169.92
InChI Key XBDUTCVQJHJTQZ-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) hydrate sulfate
SMILES O.[Fe++].[O-]S([O-])(=O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18602353
View Documents
Spectrum Chemical Mfg Cor
F1065500GM
500 g Poly Bottle
Each for $565.18
 
18602354
View Documents
Spectrum Chemical Mfg Cor
F106525KG
2.5 kg Poly Pail
Each for $1,591.03
 
18602355
View Documents
Spectrum Chemical Mfg Cor
F106512KG
12 kg Poly Pail
Each for $4,059.76
 
18602356
View Documents
Spectrum Chemical Mfg Cor
F106550KGBL
50 kg Fiber Drum
Each for $7,291.79
 
Description

Description

Spectrum™ Chemical Dried Ferrous Sulfate, Powder, USP is used to prevent or treat low levels of iron in the blood. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

13463-43-9
169.92
λ²-iron(2+) hydrate sulfate
FeH2O5S
XBDUTCVQJHJTQZ-UHFFFAOYSA-L
O.[Fe++].[O-]S([O-])(=O)=O

Specifications

13463-43-9
500 g
XBDUTCVQJHJTQZ-UHFFFAOYSA-L
λ²-iron(2+) hydrate sulfate
86 to 89%
Poly Bottle
100%
FeH2O5S
O.[Fe++].[O-]S([O-])(=O)=O
169.92
USP
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'