Drierite™ Indicating Absorbents, W.A. Hammond
Efficient desiccant giving constant visual indication of the progress of absorption and exhaustion. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
Supplier:  W A Hammond Drierite Co HDC24001
 View more versions of this product
Catalog No. 07-578-4A
Explore available promotions
Description
Description
Efficient desiccant giving constant visual indication of the progress of absorption and exhaustion. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
  • Efficient desiccant giving constant visual indication of the progress of absorption and exhaustion
  • Turns from blue to pink when exhausted
  • Dries air and gases to lower residual moisture levels (-100°F dew point)
  • Commonly used in desiccators, desiccator cabinets, in-line air and gas drying columns, absorption bulbs, U-tubes, glove boxes , sealed chambers and wherever an anhydrous environment is required
  • Regeneration by heating granules on tray in single layer at 210-220°C for 1 hour
Specifications
Specifications
Calcium Sulfate with Cobalt Chloride Indicator
Calcium Sulfate with Cobalt Chloride
98% CaSO4 with <2% CoCL2
Glass Bottle
7646-79-9,7646-79-9
1 lb.
10 to 20
Abosrbent; Desiccant; Indicating