Drierite™ Indicating Absorbents, W.A. Hammond
Efficient desiccant giving constant visual indication of the progress of absorption and exhaustion. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
Supplier: W A Hammond Drierite Co 24005
Description
Efficient desiccant giving constant visual indication of the progress of absorption and exhaustion. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
- Efficient desiccant giving constant visual indication of the progress of absorption and exhaustion
- Turns from blue to pink when exhausted
- Dries air and gases to lower residual moisture levels (-100°F dew point)
- Commonly used in desiccators, desiccator cabinets, in-line air and gas drying columns, absorption bulbs, U-tubes, glove boxes , sealed chambers and wherever an anhydrous environment is required
- Regeneration by heating granules on tray in single layer at 210-220°C for 1 hour
Specifications
Calcium Sulfate with Cobalt Chloride Indicator | |
Calcium Sulfate with Cobalt Chloride | |
98% CaSO4 with <2% CoCL2 | |
Glass Bottle |
7646-79-9,7778-18-9 | |
5 lb. | |
10 to 20 | |
Abosrbent; Desiccant; Indicating |