Drierite™ Drying Tubes, W.A. Hammond
Drying tube for air and gases. W A Hammond Drierite™ Drying Tubes dries air and gases for small scale laboratory applications and contains a flexible hose connection provides an immediate flow of dry gas.
Supplier:  W A Hammond Drierite Co 26930
Catalog No. 07-578-5
Explore available promotions
Description
Description
Drying tube for air and gases. W A Hammond Drierite™Drying Tubes dries air and gases for small scale laboratory applications and contains a flexible hose connection provides an immediate flow of dry gas.
  • Vent drier on reagent bottles, reaction apparatus, or instrument compartment,
  • For sample preparation, gravimetric determination of water vapor, drying of gases for purging and blanketing of moisture sensitive materials
  • Contains flexible hose connection, provides immediate flow of dry gas
  • Turns from blue to pink to give constant visual assurance of active desiccant
  • Polycarbonate end caps
  • 20μm polypropylene filter in each end
  • Dimensions (O.D x Length): 1.9 x 15.2cm (0.75x 6 in.)
Specifications
Specifications
Drierite Indicating Drying Tube, WA Hammond
Polycarbonate (End Caps), Polypropylene (Filter)
4 Tubes