Innovatek DriMop™ Liquid Absorbent Packets
DriMop™ Liquid Absorbent Packets absorb, and solidify up 300 times its weight of liquid
Supplier:  Innovatek 4090-02-01083CG17
Catalog No. 29-801-08
Explore available promotions
Description
Description
  • Swiftly absorbs and solidifies up to 300 times its own weight of liquid
  • Use in specimen shipping containers and specimen bags
  • Efficient containment of spills
Specifications
Specifications
0.5 g/Packet
Spill Containment
1500/Cs.