missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Droxidopa 98.0+%, TCI America™
$316.00 - $900.60
Chemical Identifiers
CAS | 23651-95-8 |
---|---|
Molecular Formula | C9H11NO5 |
Molecular Weight (g/mol) | 213.189 |
MDL Number | MFCD00799030 |
InChI Key | QXWYKJLNLSIPIN-JGVFFNPUSA-N |
Synonym | (2S,3R)-2-Amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropionic Acid, L-threo-3,4-Dihydroxyphenylserine |
PubChem CID | 92974 |
ChEBI | CHEBI:31524 |
IUPAC Name | (2S,3R)-2-amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropanoic acid |
SMILES | C1=CC(=C(C=C1C(C(C(=O)O)N)O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
D423550MG
|
TCI America
D423550MG |
50 mg |
N/A
|
|
|||||
D4235200MG
|
TCI America
D4235200MG |
200 mg |
N/A
|
|
|||||
Chemical Identifiers
23651-95-8 | |
213.189 | |
QXWYKJLNLSIPIN-JGVFFNPUSA-N | |
92974 | |
(2S,3R)-2-amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropanoic acid |
C9H11NO5 | |
MFCD00799030 | |
(2S,3R)-2-Amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropionic Acid, L-threo-3,4-Dihydroxyphenylserine | |
CHEBI:31524 | |
C1=CC(=C(C=C1C(C(C(=O)O)N)O)O)O |
Specifications
23651-95-8 | |
White-Yellow | |
MFCD00799030 | |
(2S,3R)-2-Amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropionic Acid, L-threo-3,4-Dihydroxyphenylserine | |
C1=CC(=C(C=C1C(C(C(=O)O)N)O)O)O | |
213.189 | |
CHEBI:31524 | |
≥98.0% (HPLC,T) | |
Droxidopa |
225°C | |
C9H11NO5 | |
50 mg | |
QXWYKJLNLSIPIN-JGVFFNPUSA-N | |
(2S,3R)-2-amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropanoic acid | |
92974 | |
213.19 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : VT9626010
TSCA : No