missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drummond™ Digital Microdispensers
Positive displacement piston eliminates source of errors found with air displacement types
$1679.91 - $1679.91
Specifications
| Accuracy | ±1% |
|---|---|
| Number of Channels | 1 |
| Type | Digital Microdispenser |
| For Use With (Application) | Positive lock prevents accidental volume changes |
Includes: Microdispenser 21-169-20D is supplied with 50 glass bores; 21-169-10A is supplied with 7 polypropylene bores; all other models supplied with package of 100 glass bores.
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
|
21-169-20B
|
Drummond™
3000525 |
25 μL |
Each for $1,679.91
|
|
|||||
Description
One-hand-operated positive displacement pipetter with digital reading of delivery volume to 0.01, 0.1, or 1.0μL. Five adjustable models cover volume range 0.01 to 1000μL.
- Adjustable to any volume within capacity rating
- Easy-to-set micrometer-type barrel for exact setting of volume to be pipetted
- Accuracy to ±1% at full volume
- Machined slot in barrel allows visual check for glass bore seating
- Color-coded calibration mark on bore lines up with plunger at full-capacity setting
- Borosilicate glass bores with stainless-steel plunger (PTFE tips on 25μL or larger); polypropylene bore on 1000μL size
- Use with all reagents (except hydrofluoric acid)
Order Info
Microdispenser 21-169-10A converts to a multichannel dispenser with the addition of a stainless-steel manifold.
Specifications
| ±1% | |
| 1 | |
| Digital Microdispenser | |
| Positive lock prevents accidental volume changes |