Drummond™ Original Portable Pipet-Aid™ Pipet Controller
Convenient alternative to mouth pipetting in the laboratory.
$1053.91 - $1053.91
Specifications
Autoclavable | Autoclavable |
---|---|
Type | Portable Filler/Dispenser |
For Use With (Application) | Laboratory pipetting |
For Use With (Equipment) | Glass or plastic pipets |
Includes | Pipet Controller, TC Nosepiece |
Catalog Number | Mfr. No. | Voltage | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Voltage | Price | Quantity | |||||
13-681-19
|
Drummond™
4000100 |
110 V |
Each for $1,053.91
|
|
|||||
Description
- Easy-to-use volume control mechanism permits precise fluid intake and delivery
- Check valve protects against accidental fluid intake into the handle
- Can be used with glass or plastic pipets
- Autoclavable components
Order Info
Contact your Fisher Scientific Customer Service Representative for available replacement parts.
Certifications
CE, Euro version
Specifications
Autoclavable | |
Laboratory pipetting | |
Pipet Controller, TC Nosepiece |
Portable Filler/Dispenser | |
Glass or plastic pipets |