missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Dry Field Mapping Kit
Makes the learning the concepts of electric fields easy. With the Dry Field Mapping Kit, students use the conductive paper that comes with the kit to draw any electrode configuration and then test it with an included battery.
Supplier: Eisco™ PH0918DFM
Description
- Ideal experiment for visualizing the electric field
- Easy to find the electric field around any conducting shape
- Conducive paper allows probing of equipotential lines with multimeter (not included)
- Comes with instructions
- Multi-meter and D-cell battery not included
Specifications
14 in. | |
Dry Field Mapping Kit | |
1 | |
Conductive paint, conductive and non-conductive graph paper, D-cell battery holder, corkboard, push pins, circle template, and 4 colored leads with alligator clips | |
2 kg | |
279.4 mm | |
8+ | |
Visualizing electric fields |
Mapping Kit | |
Black | |
9 to 12 | |
11 in. | |
4.6 lb. | |
355.6 mm | |
Rectangular |