Synthware™ Drying Tube, 90° Bent, Microscale
Drying tube is used to protect moisture sensitive reactions from atmosphere while permitting system to remain unsealed. For microscale applications
$39.09 - $76.41
Specifications
Product Type | 90° Bent Drying tube |
---|---|
For Use With (Application) | Organic Synthesis |
Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity | |||||
31502101
|
Synthware™
A851410 |
14/10 |
Each for $39.09
|
|
|||||
31500646
|
Synthware™
A851420 |
14/20 |
Each for $59.94
|
|
|||||
31500647
|
Synthware™
A851922 |
19/22 |
Each for $63.06
|
|
|||||
31500648
|
Synthware™
A852440 |
24/40 |
Each for $76.41
|
|
|||||
Description
- For protection of moisture sensitive reactions from atmosphere
- Joint 14/10
Specifications
90° Bent Drying tube | |
Organic Synthesis |