missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ 270 Series Sterile Frocks
Clean-processed and sterile frocks provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
Supplier: DuPont™ IC270BWHLG0030CS
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
- Garments are gamma sterilized to SAL of 10-6
- Body-length jacket design with front snap closures for easy donning and doffing
- Made of Tyvek™, a spunbonded olefin fabric that is lightweight and flexible
- Unique, patented flashspinning process creates an barrier to dry particles, microorganisms, and nonhazardous liquids
- Garments are constructed with the smooth side out to reduce particulate entrapment and bound seams reduce lint
- ISO Class 5 (formerly Class 100) compatible
- Processed in a certified ISO Class 4 (Class 10) cleanroom, antistatic treated and packaged in durable, antistatic bags
- Generous sizing permits freedom of movement
- Plastic snaps won't rust or flake during processing
- Bound seams are covered with garment fabric to reduce the potential for particle penetration
- Elastic opening for tighter fit at wrist
- Bound neck for lower particle shedding, adjustable snap at neck for better fit
- Full traceability
Specifications
Controlled Environments Frock | |
Tyvek | |
Single-use | |
Snaps | |
Bound | |
IsoClean™ 270 Series | |
91 to 96 cm | |
Clean-processed, Individually Packaged | |
104.14 cm | |
90.8 cm |
Large | |
White | |
Sterile | |
Elastic | |
None | |
36 to 38 in. | |
Long Sleeve | |
41 in. | |
35.75 in. | |
Class 100 (ISO 5) |