missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ ProShield™ 60 Coveralls, Respirator Fit Hood and Storm Flap
Coveralls with respirator fit hood are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
$218.05 - $285.85
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | Proprietary Bilaminate Composite Fabric |
Color | White |
Disposable | Single-use |
Catalog Number | Mfr. No. | Size | Height (English) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Height (English) | Price | Quantity | |||||
29-634-246
|
DuPont™
NG127SWHSM0025NP |
Small | 5 to 5.7 ft. |
Case of 25 Each for $218.09
|
|
|||||
29-634-248
|
DuPont™
NG127SWHLG0025NP |
Large | 5.5 to 5.9 ft. |
Case of 25 Each for $224.75
|
|
|||||
19-088-2161
|
DuPont™
NG127SWHXL0025NP |
X-Large | 5.8 to 6.2 ft. |
Case of 25 Each for $218.05
|
|
|||||
19-088-2162
|
DuPont™
NG127SWH2X0025NP |
2X-Large | 6 to 6.4 ft. |
Case of 25 Each for $218.09
|
|
|||||
19-088-2163
|
DuPont™
NG127SWH3X0025NP |
3X-Large | 6.2 to 6.4 ft. |
Case of 25 Each for $245.06
|
|
|||||
19-088-2164
|
DuPont™
NG127SWH4X0025NP |
4X-Large | 6.4 to 6.7 ft. |
Case of 25 Each for $273.53
|
|
|||||
19-130-270
|
DuPont™
NG127SWH5X0025NP |
5X-Large |
Case of 25 Each for $285.85
|
|
||||||
29-634-247
|
DuPont™
NG127SWHMD0025NP |
Medium | 5.3 to 5.7 ft. |
Case of 25 Each for $248.73
|
|
|||||
Description
- Microporous film that is laminated to a spundbonded polypropylene substrate.
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements.
- Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask.
- Longer zipper extends to chin for complete coverage of neck area
- Storm flap of garment material covers zipper
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Manufactured under specifications that do not contain natural rubber latex.
- Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
- ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
- Certified incinerable by independent laboratory. For details, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.
Specifications
General Purpose Coveralls | |
Proprietary Bilaminate Composite Fabric | |
Single-use | |
Elastic | |
Respirator Fit | |
Serged | |
Particle, Liquid | |
Microporous Film, Laminated to a Spundbonded Polypropylene Substrate | |
NG127S | |
25/Cs. |
Unisex | |
White | |
Elastic | |
Hooded | |
Front Zipper | |
Non-sterile | |
ASTM D751, D1117, D5034, D257, AATCC 127 | |
Yes | |
Automotive Refinishing, Waste Cleanup, Asbestos Abatement, Hazardous Remediation |