DuPont™ ProShield™ 60 Coveralls, Attached Hood and Boots
Coveralls with attached skid-resistant boots are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
Supplier:  DuPont™ NG122SWH3X002500
 View more versions of this product
Catalog No. 19-088-2149
Explore available promotions
Description
Description
  • Lightweight coveralls (1.8 oz. per sq. yd.) provide barrier protection against nontoxic particle, liquid, and spray hazards.
  • Microporous film that is laminated to a spundbonded polypropylene substrate.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements.
  • Attached hood with elastic around face opening.
  • Front zipper closure for easy donning and doffing.
  • Elastic opening for tighter fit at wrist
  • Attached ProShield 3 boots have special coating to provide added skid-resistance.
  • Manufactured under specifications that do not contain natural rubber latex.
Compliance
Meets USDA, FDA, OSHA standards; ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127 compliant.
  1. Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
  2. ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
  3. Certified incinerable by independent laboratory. For details, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic, Attached Boots
Hooded
Front Zipper
Non-sterile
USDA, FDA, OSHA, ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127
Yes
47.75 to 51.25 in.
121.28 to 130.17 cm
10 mil
37 in.
6.2 to 6.4 ft.
25/Cs.
3X-Large
Non-woven Polypropylene Substrate, Microporous Film Laminate
Single-use
Elastic
Standard, Elastic Face
Serged
Particle, Liquid
Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
NG120SWH
28.75 in.
73.02 cm
Automotive Refinishing, Waste Cleanup, Asbestos Abatement, Hazardous Remediation
93.98 cm
1.88 to 1.95 m