DuPont™ ProClean™ Limited-Use Cleanroom Frocks
Maximum containment short of a full coverage coverall. DuPont™ ProClean™ limited-use, full-length frocks offer the breathability and comfort of microporous film fabric.
Supplier:  DuPont™ PC271SWHLG00300B
 View more versions of this product
Catalog No. 19-0881019
Explore available promotions
Description
Description
Proprietary polypropylene laminate fabric provides effective contamination control and protection from liquid-splash and dry-particulate cleanroom hazards.
  • White knee-length frocks
  • Microporous film fabric
  • Serged seams
  • Elastic wrist
  • Bound neck
  • Available with a five-snap or zipper front closure
  • Antistatic treated
Specifications
Specifications
Controlled Environments Frock
Microporous Composite Fabric
Non-sterile
Elastic
Class 1000 (ISO 6)
Large
White
Zipper
Serged