DuPont™ ProShield™ 10 Coveralls, Attached Hood and Skid-Resistant Boots
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
Supplier:  DuPont™ PB122SWHMD002500
 View more versions of this product
Catalog No. 19162514
Explore available promotions
Description
Description
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
  • Comfort fit design based on extensive wearer input provides a comfortable garment design that enables a greater range of movement while stretching and bending, a more tailored fit, reinforcement in high stress areas for fewer blowouts, a longer zipper for easier donning and doffing and an elastic waist to better position the garment
  • Attached hood with elastic around face opening
  • Elastic opening for tighter fit at wrist
  • Attached skid-resistant boots
  • White or blue
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Elastic
Attached
Non-sterile
PB122S
25/Cs.
Medium
SMS Fabric
Single-use
Attached Boots
Hooded
Front Zipper
ASTM D1117
General Maintenance, Janitorial, Cleaning