missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ ProShield™ 10 Coveralls, Attached Hood and Skid-Resistant Boots
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
Supplier: DuPont™ PB122SBUXL002500
Description
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
- Comfort fit design based on extensive wearer input provides a comfortable garment design that enables a greater range of movement while stretching and bending, a more tailored fit, reinforcement in high stress areas for fewer blowouts, a longer zipper for easier donning and doffing and an elastic waist to better position the garment
- Attached hood with elastic around face opening
- Elastic opening for tighter fit at wrist
- Attached skid-resistant boots
- White or blue
Specifications
General Purpose Coveralls | |
Unisex | |
Blue | |
Elastic | |
Elastic | |
Attached | |
Non-sterile | |
PB122S | |
25/Cs. |
X-Large | |
SMS Fabric | |
Single-use | |
Attached Boots | |
Hooded | |
Front Zipper | |
ASTM D1117 | |
General Maintenance, Janitorial, Cleaning |