missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ ProShield™ 10 Hooded Coveralls
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
Supplier: DuPont™ PB127SWHMD002500
Description
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
- Spunbond meltblown spunbond (SMS) coveralls have been optimized for comfort, softness, and breathability.
- Designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications.
- Applications include: general maintenance, janitorial/cleaning and other dirty work.
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements.
- Attached hood with elastic around face opening.
- Elastic opening for tighter fit at wrist.
- Elastic opening for tighter fit at ankle.
- Manufactured under specifications that do not contain natural rubber latex.
Specifications
General Purpose Coveralls | |
Unisex | |
White | |
Elastic | |
Hooded | |
Front Zipper | |
Non-sterile | |
Yes | |
General Maintenance, Janitorial, Cleaning |
Medium | |
SMS Fabric | |
Single-use | |
Elastic | |
Attached | |
Serged | |
Particle | |
PB127SWH | |
25/Cs. |