DuPont™ ProShield™ 10 Hooded Coveralls
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
Supplier:  DuPont™ PB127SBULG002500
 View more versions of this product
Catalog No. 19-162-550
Explore available promotions
Description
Description
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
  • Spunbond meltblown spunbond (SMS) coveralls have been optimized for comfort, softness, and breathability.
  • Designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications.
  • Applications include: general maintenance, janitorial/cleaning and other dirty work.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements.
  • Attached hood with elastic around face opening.
  • Elastic opening for tighter fit at wrist.
  • Elastic opening for tighter fit at ankle.
  • Manufactured under specifications that do not contain natural rubber latex.
Specifications
Specifications
General Purpose Coveralls
Unisex
Blue
Elastic
Hooded
Front Zipper
Non-sterile
Yes
General Maintenance, Janitorial, Cleaning
Large
SMS Fabric
Single-use
Elastic
Attached
Serged
Particle
PB127SWH
25/Cs.