DuPont™ ProShield™ 10 Coveralls, Elastic Wrists and Ankles
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
Supplier:  DuPont™ PB125SWH3X002500
 View more versions of this product
Catalog No. 19-162-532
Explore available promotions
Description
Description
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
  • Soft, breathable and lightweight with comfort fit design
  • Spunbond Meltblown Spunbond (SMS)
  • Disposable
  • Laydown collar
  • Elastic wrists, ankles
  • Front zipper closure
  • Serged seams
  • Available in blue or white
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Elastic
Unhooded
Non-sterile
PB125S
25/Cs.
3X-Large
SMS Fabric
Single-use
Elastic
Collared
Front Zipper
Laydown
General Maintenance, Janitorial, Cleaning