missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Proshield™ 10 Frocks, Enclosed Elastic Wrists
SMS frocks are designed to be used in a range of industries for nonhazardous dry particle and light splash applications.
$128.87 - $165.72
Specifications
Product Type | General Purpose Frock |
---|---|
Material | SMS |
Color | White |
Disposable | Single-use |
Closure Type | Snaps |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-162-635
|
DuPont™
PB267SWHMD003000 |
Medium |
Case of 30 Each for $136.59
|
|
|||||
19-162-636
|
DuPont™
PB267SWHLG003000 |
Large |
Case of 30 Each for $128.87
|
|
|||||
19-162-637
|
DuPont™
PB267SWHXL003000 |
X-Large |
Case of 30 Each for $136.59
|
|
|||||
19-162-639
|
DuPont™
PB267SWH3X003000 |
3X-Large |
Case of 30 Each for $144.79
|
|
|||||
19-162-638
|
DuPont™
PB267SWH2X003000 |
2X-Large |
Case of 30 Each for $144.79
|
|
|||||
19-162-640
|
DuPont™
PB267SWH4X003000 |
4X-Large |
Case of 30 Each for $165.72
|
|
|||||
Description
- Disposable frocks that are optimized for comfort, softness and breathability
- Spunbond meltblown spunbond (SMS) material
- Snap closure (six plus one adjustable)
- Enclosed elastic wrists
- Mandarin collar
- Serged seams
- Set sleeves
Specifications
General Purpose Frock | |
White | |
Snaps | |
Serged | |
Mandarin |
SMS | |
Single-use | |
Elastic | |
General Maintenance, Janitorial, Cleaning | |
Yes |