DuPont™ Tychem™ CPF™ 3 HD Personal-Protection Garments
Integrated gloves eliminate need for taping and provide improved protection
Supplier:  DuPont™ C3184TTN3X000600
 View more versions of this product
Catalog No. 19-130-1024F
Explore available promotions
Description
Description
Garments deliver proven barrier protection against 110 toxic industrial chemicals and are designed to stand up to chemical warfare agents including Sarin, Mustard, and Lewisite † Choose from two styles: with three-panel respirator-fitted hood or with collar for compatibility with hooded PAPR.
  • Tactile glove integrated into garment†—Stretch Chloroprene with Ansell™ Barrier™ inner glove for wide spectrum chemical protection
  • Heavy-duty front zipper closure and double storm flaps with wide hook and loop closure promote improved performance
  • Bigger zipper pull ensures easier donning and doffing
  • Includes attached socks‡ with boot splash cover
  • Taped seams
  • Light tan
Order Info
Basis Weight: 4.5 oz./yd.2 per ASTM™ D751. Ball Burst (md/cd): 47lbf per ASTM D750. Grab Tensile Strength (md/cd): 60/50 ft.-lb per ASTM D751.
†Review separate permeation data on both gloves and the fabric before selecting an ensemble.
‡Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.
Specifications
Specifications
3X-Large
Tan
Attached Gloves
Front Zipper with Storm Flap
Collar, attached dual layer gloves, attached socks with outer boot flaps; Multilayer film barrier laminated to Polypropylene substrate
31 in.
78.74 cm
40.5 in.
31.75 in.
1.88 to 1.95 m
Tychem
Attached Socks, Boot Overflaps
Collared
Taped
50.25 to 53.75 in.
127.63 to 136.52 cm
Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
102.87 cm
6.2 to 6.4 ft.