missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tychem™ CPF 3 Hoods
DuPont Tychem 5000 Hood. Standard Visor, 1 Layer: 20 mil PVC. Extends Over Chest. Hook and Loop Waist Closure. Taped Seams. Tan.
Supplier: DuPont™ C3651TTN00000600
Description
Tychem 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate. Tychem 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist.Extends over shoulders to provide full head protectionTaped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.20 mil PVC visorAttached vest design with elastic at shoulders and strap for torso to secure in place.Specifications
Hood with Visor | |
Tychem | |
Tan | |
2 oz. |