DuPont™ Tychem™ Reflector™ Encapsulated Level A Suit
Encapsulated Level A garment design for highest level of protection from liquid splash and vapor/gas exposures for both wearer and respiratory equipment
Supplier:  DuPont™ RF600TSV3X000100
Catalog No. 19-067-029
Explore available promotions
Description
Description
Tychem™ Reflector™ garments are composed of a patented, heavy-duty aluminized fabric made of a Nomex™/DuPont™ Kevlar™ blend laminated to a multi-layer barrier film. This unique combination of layers provides excellent physical strength and cut and abrasion protection. Ideal for industrial chemical, HazMat response and military applications including chemical warfare agents.
  • Triple taped seams provide strong chemical resistance against heavy liquid splashes
  • Sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing
  • Standard width, four layer faceshield
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Vapor tight zipper is extra long to increase garment opening and aid donning and doffing
  • Storm flap covers zipper which can be secured by wearer with rugged hook and loop material to prevent intrusion at zipper
  • Double storm flap with hoop and loop closure
  • Three component glove system: inner North Silver Shield™ / 4H™ glove provides broad range chemical protection, middle Neoprene gloves provide mechanical and physical durability to protect inner SilverShield™ glove and outer knit glove made of Kevlar™ for abrasion and cut resistance
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment
  • One-way valves are positive pressure and open on demand
  • Valves include splash covers made of barrier material to help prevent liquid intrusion
  • Valves are located on left back of head and right lower back
  • Internal adjustable waist belt system for support and improved fit
  • Each suit has unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
Certifications
NFPA 1991 (2005 edition) certified
Specifications
Specifications
Silver
3X-Large
33 in.
Tychem
6 ft. 3 in. to 6 ft. 5 in.