Learn More
DuPont™ Tychem™ 4000 Hooded Coverall
Supplier: DuPont™ SL127TWH5X000600
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
- Attached hood with elastic around face opening.
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Manufactured under specifications that do not contain natural rubber latex
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Specifications
| 5X-Large | |
| Tychem | |
| Elastic | |
| Attached | |
| Taped | |
| Waste management, hazardous response and nuclear,environments |
| Unisex | |
| White | |
| Hooded | |
| Front Zipper | |
| Attached hood |