missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tychem™ SL 275 Series Aprons
$540.79 - $656.26
Specifications
Material | Tychem |
---|---|
Color | White |
For Use With (Application) | Waste management, hazardous response and nuclear environments |
Length (English) | 44 in. |
Length (Metric) | 111.7 cm |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-162-788
|
DuPont™
SL275TWH4X002500 |
4X-Large |
Case of 25 Each for $656.26
|
|
|||||
19-162-789
|
DuPont™
SL275TWHSM002500 |
Small |
Case of 25 Each for $540.79
|
|
|||||
Description
DuPont Tychem 4000 delivers effective protection against a range of chemical environments. Tychem 4000, utilizes Saranex 23-P film laminated to Tyvek fabric to provide a rugged and durable fabric that resists abrasion. Tychem 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.Knee-length apron/gown design with attached sleevesProvides enhanced frontal protectionTaped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.Open back for wearer comfortBound seams around neck opening with snap closureTies at waistElastic opening for tighter fit at wrist44 in. long for neck-to-knee coverageAvailable in multiple sizesSpecifications
Tychem | |
Waste management, hazardous response and nuclear environments | |
111.7 cm |
White | |
44 in. | |
Taped |