Learn More
DuPont™ Tychem™ ThermoPro™ Coveralls
Provides chemical, flash fire and electric arc protection
$1446.27 - $1754.99
Specifications
Material | Tychem |
---|---|
Ankle Style | Attached Socks, Boot Overflaps |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | Respirator Fit (Drawstring) |
Alert:
Do not use for protection against continuous thermal loads, molten metal splash, hot liquids or steam. Do not wear for structural or proximity fire fighting.
Catalog Number | Mfr. No. | Color | Material | Size | Certifications/Compliance | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Material | Size | Certifications/Compliance | Price | Quantity | ||
19-101-346
|
DuPont™
TP199TOR3X000200 |
Orange | Tychem | 3X-Large | NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ |
Case of 2 Each for $1,625.03
|
|
||
19-101-344
|
DuPont™
TP199TOR2X000200 |
Orange | Tychem | 2X-Large | NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ |
Case of 2 Each for $1,533.03
|
|
||
19-101-348
|
DuPont™
TP199TOR4X000200 |
Orange | Tychem | 4X-Large | NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ |
Case of 2 Each for $1,754.99
|
|
||
19-101-351
|
DuPont™
TP199TORLG000200 |
Orange | Tychem | Large | NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ |
Case of 2 Each for $1,446.27
|
|
||
19-101-353
|
DuPont™
TP199TORMD000200 |
Orange | Tychem | Medium | NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ |
Case of 2 Each for $1,446.27
|
|
||
19-101-356
|
DuPont™
TP199TORXL000200 |
Orange | Tychem | X-Large | NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ |
Case of 2 Each for $1,446.27
|
|
||
Description
Coveralls combine the chemical protection of DuPont Tychem with the flame and arc flash protection of DuPont Nomex™ into a single garment. The fabric delivers permeation protection against a broad range of toxic industrial chemicals and even chemical warfare agents.
- Provide at least eights hours barrier to 20 of the 21 chemicals found in ASTM™ F1001
- Attached respirator-fit hood with drawstring
- Zipper front with Velcro™ fastener double storm flaps fastened by flame-resistant treated polyamide hook-and-loop material
- Elastic at the wrists and ankles
- Attached socks and boot flaps
- Available in high-contrast orange or low-visibility gray
- Arc rating:15.2 cal/cm2†
- Thickness: 34mil
- Basis Weight: 841 oz./yd.2
- Ball Burst: 167 lbf
- Tear Resistance–Trap Tear (MD/CD): 25/32 lbf
- Breaking Strength–Grab (MD/CD): 155/170 lbf
- Wearing Apparel Flammability: Class 1
† For electric arc hazard exposures, wear the proper arc rated garment system as dictated by an electric arc hazard analysis.
Specifications
Tychem | |
Elastic | |
Respirator Fit (Drawstring) | |
Taped | |
Respirator fit, drawstring hood, elastic wrist, attached socks with outer boot flaps, double storm flap with hook and loop closure | |
Fire departments, industrial fire brigades, clandestine lab investigation, industrial chemical processing plants | |
0.23 kg |
Attached Socks, Boot Overflaps | |
Hooded | |
Zipper with Double Storm Flap, Hook and Loop Seal | |
15.2 Calories/cm2 | |
34 mil | |
0.51 lb. |