missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tychem™ ThermoPro™ TP198T Coveralls
ThermoPro coveralls offer superior protection against fire, electricity, and toxic chemicals.
$1234.66 - $1618.14
Specifications
Material | HDPE |
---|---|
Ankle Style | Open |
Cuff Style | Elastic |
Neck Style | Hooded |
Closure Type | Hook and Loop |
Catalog Number | Mfr. No. | Certifications/Compliance | Color | Size | Height (English) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | Color | Size | Height (English) | Price | Quantity | ||
19-101-316
|
DuPont™
TP198TOR3X000200 |
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ | Orange | 3X-Large | 6.2 to 6.4 ft. |
Case of 2 Each for $1,387.28
|
|
||
19-101-318
|
DuPont™
TP198TOR4X000200 |
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ | Orange | 4X-Large | 6.4 to 6.7 ft. |
Case of 2 Each for $1,498.28
|
|
||
19-101-320
|
DuPont™
TP198TOR5X000200 |
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ | Orange | 5X-Large | 6.7 to 6.10 ft. |
Case of 2 Each for $1,618.14
|
|
||
19-101-322
|
DuPont™
TP198TORLG000200 |
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ | Orange | Large | 5.5 to 5.9 ft. |
Case of 2 Each for $1,234.66
|
|
||
19-101-324
|
DuPont™
TP198TORMD000200 |
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ | Orange | Medium | 5.3 to 5.7 ft. |
Case of 2 Each for $1,234.66
|
|
||
19-101-327
|
DuPont™
TP198TORXL000200 |
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ | Orange | X-Large | 5.8 to 6.2 ft. |
Case of 2 Each for $1,234.66
|
|
||
Description
ThermoPro coveralls are secure in fit to seal out contaminants
- Respirator fit hood with drawstrings
- Front zipper with storm flap, hook and loop secure
- Elastic wrists
- Open, hemmed ankles
- Provides at least one hour breakthrough protection against 20 of the 21 standard ASTM F1001 chemicals
- At least 30 minutes of protection against 182 chemical challenges
- Protects against flash fire, electric arc and liquid-chemical splash, wide range of toxic and chemical warfare agents
- HazMat teams in fire departments
- Industrial fire brigades
- Clandestine lab investigation
- Industrial chemical processing plants
Certifications
- NFPA 1992 (2012)
- NFPA 2112 (2012)
- Exceeds NFPA 70E Hazard Risk Category 2
Specifications
HDPE | |
Elastic | |
Hook and Loop | |
Fit hood, elastic wrist, open ankle, with respirator; HDPE Tychem™ ThermoPro construction |
Open | |
Hooded | |
Taped | |
Fire departments, industrial fire brigades, clandestine lab investigation, industrial chemical processing plants |