DuPont™ Tyvek™ Bib Apron
Nonwoven Tyvek fabric helps resist penetration by particles and other contaminants. The inherent barrier stops submicron particles even after abrasion.
Supplier:  DuPont™ TY273BWH00010000
Catalog No. 01-361-83
Explore available promotions
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
Nonwoven Tyvek fabric helps resist penetration by particles and other contaminants. The inherent barrier stops submicron particles even after abrasion.
  • Provides enhanced frontal protection
  • Bound seams around neck
  • Ties at waist
  • One size fits most
Specifications
Specifications
Bib Apron
Tyvek
Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up
69 cm
Universal
White
36 in.
Bound