missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Series 264 Frocks, Clean-Processed and Sterile
Sterile and clean-processed frocks offer lowest level of particle shedding within the DuPont portfolio. Tyvek™ provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
$610.48 - $740.85
Specifications
Product Type | Controlled Environments Frock |
---|---|
Material | Tyvek |
Color | White |
Disposable | Single-use |
Sterility | Sterile |
Alert:
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog Number | Mfr. No. | Size | Length (English) | Length (English) Sleeve | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Length (English) | Length (English) Sleeve | Price | Quantity | |||
17-990-513
|
DuPont™
IC264SWHSM0030CS |
Small | 41 in. | 34 in. |
Case of 30 Each for $610.48
|
|
|||
17-990-514
|
DuPont™
IC264SWHMD0030CS |
Medium | 42.75 in. | 35.5 in. |
Case of 30 Each for $610.48
|
|
|||
17-990-515
|
DuPont™
IC264SWHLG0030CS |
Large | 45 in. | 37.5 in. |
Case of 30 Each for $610.48
|
|
|||
17-990-516
|
DuPont™
IC264SWHXL0030CS |
X-Large | 47 in. | 39.25 in. |
Case of 30 Each for $610.48
|
|
|||
17-990-517
|
DuPont™
IC264SWH2X0030CS |
2X-Large | 53 in. | 42.75 in. |
Case of 30 Each for $647.19
|
|
|||
17-990-518
|
DuPont™
IC264SWH3X0030CS |
3X-Large | 55 in. | 44.25 in. |
Case of 30 Each for $685.93
|
|
|||
17-990-519
|
DuPont™
IC264SWH4X0030CS |
4X-Large | 57 in. | 46 in. |
Case of 30 Each for $740.85
|
|
|||
Description
- Frocks are gamma sterilized to SAL of 10-6
- Clean-processed
- Serged seams have multiple interlocking threads sewn around the raw edges
- Elastic openings for tighter fit at wrist
- Bound neck for lower particle shedding
- Front zipper closure for easy donning and doffing
- Raglan sleeve design for greater range of motion and comfort
- A-line silhouette
- Individually packaged, 30 per case
Specifications
Controlled Environments Frock | |
White | |
Sterile | |
Elastic | |
Bound | |
Raglan Sleeve |
Tyvek | |
Single-use | |
Zipper | |
Serged | |
IsoClean™ 264 Series | |
Clean-processed, Individually Packaged |