DuPont™ Tyvek™ IC668B IsoClean™ Full Face Opening Hoods
IsoClean open face hoods provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Available in non-sterile, clean-processed, and sterile versions.
Supplier:  DuPont™ IC668BWH000100CS
Catalog No. 17988113
Explore available promotions
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
  • Bound seams are covered with garment fabric for reinforcement
  • Bound hood opening for lower particle shedding
  • Full face opening
  • Ties with loops for adjustable fit
  • One size fits most
  • Availiable in non-sterile, clean-processed, and sterile versions
  • Available in bulk packaging with double transparent poly liners
  • Clean-processed garments meet ISO Class Cleanroom 4 standards
  • Sterilzed hoods = gamma sterilzed to SAL of 10-6
Specifications
Specifications
Full Face Opening
Tyvek
Ties
IC668B
Sterile
Ties with loops for fit
Class 1-10 (ISO 3-4)
Universal
White
Single-use
Bound
100/Cs.
Clean and Sterile