DuPont™ Tyvek™ IsoClean™ Series 447 Boot Covers
Gripper™ soles resists slipping on slick surfaces
Supplier:  DuPont™ IC447SWHMD0100CS
 View more versions of this product
Catalog No. 19-039-851
Explore available promotions
Description
Description
Tyvek IsoClean fabric is antistatic and low linting and provides an excellent barrir to dry particulates, nonhazardous liquids and microorganisms.
  • Coarsely textured interior to help prevent movement inside
  • PVC Gripper soles provide excellent barrier to liquid penetration
  • 18 in. (45.7cm) high Tyvek uppers with elastic leg and ankle openings
  • Serged seams
  • White
  • Available clean processed and gamma sterilized or bulk packaged
Order Info
Clean and sterile covers are individually packaged in opaque bag and placed in double transparent poly liners in and ISO Class 4 cleanroom. Bulk covers are bulk packaged in double transparent poly liners.
Specifications
Specifications
Medium
Yes
Sterile
Serged
Ideal to provide an excellent barrier to dry particles, nonhazardous liquids and microorganisms
27.3 cm
45.7 cm
Tyvek Flashspun Polyethylene
White
Elastic Top and Ankles
50 pairs
10.75 in.
18 in.
Product Suggestions
Product Suggestions