DuPont™ Tyvek™ IsoClean™ IC461 Shoe Covers
Serged seam shoe covers feature an anti-skid PVC sole and are suitable for ISO 6 (Class 1000) controlled environments.
Supplier:  DuPont™ IC461SWHXL03000B
 View more versions of this product
Catalog No. 17-988-106
Explore available promotions
Description
Description
  • Serged seams
  • Elastic opening
  • Elastic toe
  • PVC sole
  • 5 in. (12.7cm) high
  • Bulk-packaged
  • White
Specifications
Specifications
X-Large
Yes
Single-use
Anti-skid
5.4 mil
15.75 in.
5.25 in.
Class 1000 (ISO 6)
Tyvek Flashspun Polyethylene
White
Elastic Ankle
Serged
ASTM D3776, ASTM F2101, ASTM D774, ASTM D5034, ASTM D257, AATCC 127, 16 CFR 1610 Class 1
40 cm
13.33 cm