missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Series 253 Coveralls, Clean-Processed and Sterile
Clean-proceesed and sterile coveralls worn in controlled environments protect against particulates and other contaminants.
Supplier: DuPont™ IC253BWH2X0025CS
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Clean-proceesed and sterile coveralls worn in controlled environments protect against particulates and other contaminants.
- Garments are gamma sterilized to SAL of 10-6
- Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Coverall has elastic openings for tighter fit at wrist and ankle
- Bound neck for lower particle shedding
- Front zipper closure for easy donning and doffing
- Dolman sleeve design for greater range of motion and comfort
- Full traceability on all sterilized apparel with certificates of sterility available
Specifications
Cleanroom Coveralls | |
Tyvek™ | |
Covered Elastic | |
Collared | |
Bound | |
Bound | |
43 to 46 in. | |
109.2 to 116.84 cm | |
Dolman | |
Barrier to dry particles, microorganisms, and non-hazardous liquids | |
100.96 cm | |
185.92 to 198.12 cm |
2X-Large | |
White | |
Covered Elastic | |
Zipper | |
Sterile | |
IC253BWH | |
34 in. | |
86.3 cm | |
5 mil | |
39.75 in. | |
6.1 to 6.5 ft. |