DuPont™ Tyvek™ 400 Pullover Hood
Tyvek spunbound olefin hood provides protection against microscopic airborne particles.
Supplier:  DuPont™ TY657SWH00010000
Catalog No. 01-361-75D
Explore available promotions
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
  • Lightweight, chemically and biologically inert and tear resistant
  • Features tunnelized elasticized face opening and provides complete coverage of head and shoulders
  • Excellent for use with coveralls
  • One-size-fits-all
  • White
Order Info
Case of 100.
Specifications
Specifications
Overhood
Tyvek