H-B Instrument™ Durac™ Bi-Metallic Dial Thermometers
Features 1/2 in. NPT threaded connection and polycarbonate lens.
$76.77 - $161.63
Specifications
For Use With (Application) Petrochemical, oil and gas, waste water, pharmaceutical, compost, food and beverage storage and processing, asphalt, mining, utilities, automotive, marine, photographic
Lens Material Polycarbonate
Material Metal
Accuracy 0.01
View More Specs
Products 2
Catalog Number Mfr. No. Temperature Range (Metric) Temperature Range (English) Dial Size (Metric) Length (Metric) Probe Price Quantity  
Catalog Number Mfr. No. Temperature Range (Metric) Temperature Range (English) Dial Size (Metric) Length (Metric) Probe Price Quantity  
S02684
View Documents
H-B Instrument™
B613107200
-20°C to +120°C 0°F to +250°F 75 mm 101.6 mm
Each for $161.63
S02668
View Documents
H-B Instrument™
B613105700
-10°C to +110°C 0°F to +220°F 50 mm 200 mm
Each for $76.77
Description
Description
  • External adjustment allows for simple field calibration
  • For correct temperature reading, stem must be immersed at least 50mm in liquids and 100mm in gases
  • Each thermometer is individually packaged with a multi-language (English, French, German, Italian, Portuguese and Spanish) Statement of Test and Calibration
  • Part No 21596 features 3/8 in. NPT threaded connection which is ideal for industrial installations in tank or pipe lines
  • 1.7 in. dial size thermometers (except Part No 21596) do not have NPT threaded connections
  • They feature a beaker clip with built in recalibration adjustment tool
Specifications
Specifications
Petrochemical, oil and gas, waste water, pharmaceutical, compost, food and beverage storage and processing, asphalt, mining, utilities, automotive, marine, photographic
Polycarbonate
Metal
0.01
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'