missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences DURAN™ GL 45 Screw Caps for Laboratory Glass Bottles
Distinguishing media types is simplified and interchanging of screw caps and carry over of substances is effectively excluded.
$95.26 - $370.29
Specifications
Product Type | Screw Cap |
---|---|
For Use With (Equipment) | Laboratory glass bottles |
Closure Size | GL 45 |
Catalog Number | Mfr. No. | Color | Material | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Material | Price | Quantity | ||||
09-841-270
|
DWK Life Sciences
292392809 |
Blue | Polypropylene |
Pack of 10 Each for $95.26
|
|
||||
09-841-278
|
DWK Life Sciences
293382884 |
Gray | Polypropylene |
Pack of 10 Each for $95.26
|
|
||||
09-841-277
|
DWK Life Sciences
293382868 |
Green | Polypropylene |
Pack of 10 Each for $95.26
|
|
||||
09-841-271
|
DWK Life Sciences
292402807 |
Red | PBT |
Pack of 10 Each for $370.29
|
|
||||
09-841-279
|
DWK Life Sciences
293382802 |
Yellow | Polypropylene |
Pack of 10 Each for $95.26
|
|
||||
Description
- Available in blue, green, yellow and grey with match pour rings
Specifications
Screw Cap | |
Laboratory glass bottles | |
GL 45 |